Learn More
IBA Lifesciences CD3 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™
Brand:  IBA Lifesciences 6-8001-150
Additional Details : Weight : 0.00600kg
 View more versions of this product
Product Code. 17217600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • CD3 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
  • Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
  • Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells
  • Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology.
Specifications
Specifications
Lyophilized
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation
Human
Human CD3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only.