Learn More
Abnova™ CD273(PDCD1LG2)/CEN9q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0225.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16073365
  • 10615.00 DKK / 100µL
Estimated Shipment: 01-08-2023
to see stock.
Description
Description
Specifications
Specifications
80380
B7DC,Btdc,CD273,MGC142238,MGC142240,PD-L2,PDCD1L2,PDL2,bA574F11.2
Fluorescent In Situ Hybridization (Cell)
100 μL
PDCD1LG2
Store at 4°C in the dark
programmed cell death 1 ligand 2
DNA/RNA
Liquid