Learn More
Abnova™ CD273(PDCD1LG2)/CEN9q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
11735.00 DKK - 19325.00 DKK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | programmed cell death 1 ligand 2 |
Gene Alias | B7DC,Btdc,CD273,MGC142238,MGC142240,PD-L2,PDCD1L2,PDL2,bA574F11.2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073365
|
Abnova™
FG0225.100UL |
100 μL |
11735.00 DKK
100µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083365
|
Abnova™
FG0225.200UL |
200 μL |
19325.00 DKK
200µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
B7DC,Btdc,CD273,MGC142238,MGC142240,PD-L2,PDCD1L2,PDL2,bA574F11.2 | |
80380 | |
Liquid |
Tube | |
programmed cell death 1 ligand 2 | |
PDCD1LG2 | |
DNA/RNA |