missing translation for 'onlineSavingsMsg'
Learn More

CD25 Mouse anti-Human, Super Bright 600, Clone: BC96, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen 63-0259-41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15855563

  • 1124.00 DKK / Pack of 25


Estimated Shipment date
from Supplier 29-05-2019
Add to basket

Description

Description

Light sensitivity: This tandem dye is sensitive to photo- induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 2-8°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorochrome performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
Specifications

Specifications

CD25
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P01589
Interleukin-2 Receptor alpha, IL-2Ra
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BC96
Super Bright 600
IL2RA
Liquid
IL2RA
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3559
Videos

For Research Use Only