missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD238 Mouse anti-Human, BUV737, Clone: BRIC 203, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748631
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD238 Monoclonal antibody specifically detects CD238 in Human samples. It is validated for Flow Cytometry
Tekniske data
| CD238 | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| KEL; KELL; Kell blood group, metallo-endopeptidase; ECE3 | |
| Human erythrocytes | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| BRIC 203 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion