Learn More
CD235a (Glycophorin A) Chimeric Recombinant Rabbit Monoclonal Antibody (M2A1), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA548073
Description
Specificity: M2A1 binds specifically to the M antigen on human erythrocytes, and shows no reactivity to N antigen. The epitope for this antibody includes the amino-terminal serine residue and sialic acid residues of glycophorin A. The reactivity of M2A1 for M antigen is pH- and salt-dependent (optimum ∽ pH8-9). Blood group antigen M is located on the major sialoglycoprotein (glycophorin A) of red blood cells.
CD235a (Glycophorins A, GPA) is a single pass membrane sialoglycoprotein expressed in mature erythrocytes and erythroid precursor cells. CD235a is the carrier of blood group M and N specificities. CD235a provides cells with a large mucin-like that may serve as a barrier to cell fusion, minimizing aggregation between red blood cells in the circulation. CD235a has been shown to act as a receptor for Sandei virus, parvovirus, and Hsa, an Streptococcus adhesin.
Specifications
CD235a (Glycophorin A) Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P02724 | |
GYPA | |
M2A1 was prepared by immunzing BALB/c mice with human red blood cells of the M phenotype, and was screened by testing for anti-M activity. | |
200 μg | |
Primary | |
Human | |
Liquid |
ELISA, Inhibition Assays, Western Blot | |
M2A1 | |
Unconjugated | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Rabbit | |
Affinity chromatography | |
RUO | |
2993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.