missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), NovaFluor™ Blue 585, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1155.00 DKK - 2562.00 DKK
Tekniske data
| Antigen | CD22 |
|---|---|
| Klon | eBio4KB128 (4KB128) |
| Koncentration | 0.2 μg/Test |
| Applikationer | Flow Cytometry |
| Klassifikation | Monoclonal |
Beskrivelse
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow CytometryTekniske data
| CD22 | |
| 0.2 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| eBio4KB128 (4KB128) | |
| Flow Cytometry | |
| NovaFluor Blue 585 | |
| Mouse | |
| Human | |
| P20273 | |
| 933 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| CD22 |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel