missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R4 Antibody (4), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90661G
Additional Details : Weight : 0.00970kg
Description
CD200R4 Monoclonal specifically detects CD200R4 in Mouse samples. It is validated for ELISA.Specifications
CD200R4 | |
Monoclonal | |
DyLight 488 | |
F630107N04Rik, MCD200, MCD200RLa | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R4 (NP_997127.1; Met 1-Thr 241). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
4 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
239849 | |
Store at 4°C in the dark. | |
IgG |