missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD200/OX2 Antibody (samalizumab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28596JF635
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD200/OX2 Monoclonal antibody specifically detects CD200/OX2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
CD200/OX2 | |
Monoclonal | |
Janelia Fluor 635 | |
antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
CD200 | |
0.1 mL | |
Immunology | |
4345 | |
Store at 4°C in the dark. | |
IgG2 |
Flow Cytometry, ELISA, Functional Assay | |
samalizumab | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
CD200/OX2 Antibody (samalizumab) [Janelia Fluor™ 635], Novus Biologicals™ > 0.1 mL; Janelia Fluor 635
Ser du en mulighed for forbedring?Del en indholdskorrektion