Learn More
CD152 (CTLA-4) Monoclonal Antibody (14D3), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
594.00 DKK - 1394.00 DKK
Specifications
Antigen | CD152 (CTLA-4) |
---|---|
Clone | 14D3 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Functional Assay, Neutralization |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15207257
|
Affymetrix eBioscience
14-1529-82 |
100 μg |
1394.00 DKK
100µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15297247
|
Affymetrix eBioscience
14-1529-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 14D3 monoclonal antibody reacts with human CD152, also known as cytotoxic T lymphocyte antigen-4 (CTLA-4). CTLA-4, a protein with structural similarities to CD28, is expressed on activated T cells (activated B cells may also express CTLA-4) and binds the B7 family members, CD80 (B7-1) and CD86 (B7-2), with higher affinity than CD28 does. CTLA-4 and CD28 appear to deliver opposing signals to T cells: while CD28 is a potent costimulator, CTLA-4 restricts the progression of T cells to an activated state by inhibiting IL-2 secretion and cellular proliferation. The cytoplasmic portion of CTLA-4 contains ER retention motifs, resulting in intracellular localization of a large proportion of newly synthesized CTLA-4 in response to TCR signaling. The 14D3 antibody also recognizes rhesus monkey and has inhibitor activity. Applications Reported: The 14D3 antibody has been reported for use in flow cytometric analysis and immunoblotting (WB). 14D3 has also been reported in in vitro inhibition studies. (Please use Functional Grade purified 14D3, cat. 16-1529, in functional assays.). Applications Tested: The 14D3 antibody has been tested by intracellular staining and flow cytometric analysis of PHA stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol.
CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.Specifications
CD152 (CTLA-4) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P16410 | |
CTLA4 | |
Primary | |
4° C | |
CTLA4 |
14D3 | |
Flow Cytometry, Functional Assay, Neutralization | |
Unconjugated | |
Mouse | |
Human, Rhesus Monkey | |
1493, 705673 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.