Få mere at vide
Novus Biologicals™ CD133 Recombinant Protein Antigen
Artikelnummer. 18245412 Shop alle Bio Techne produkter
Skift visning
Protein længde:
LQDFAACGIDRMNYDSYLAQTGKSPAGVNLLSFAYDLEKANSLPPGNLRNSLKRDAQTIKTIHQQRVLPIEQSLSTLYQSVKILQRTGNGLLERVTRILASLDFAQNFITNNTSSVIIEETTKKYGRTII
QYNTTKDKAFTDLNSINSVLGGGILDRLRPNIIPVLDEIKSMATAIKETKEALENMNSTLKSLHQQSTQLSSSLTSVKTSLRSSLNDPLCLVHPSSETCNSIRLSLS
Pakningsstørrelse:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde unitSize
18245412 LQDFAACGIDRMNYDSYLAQTGKSPAGVNLLSFAYDLEKANSLPPGNLRNSLKRDAQTIKTIHQQRVLPIEQSLSTLYQSVKILQRTGNGLLERVTRILASLDFAQNFITNNTSSVIIEETTKKYGRTII 0.1ml
18253932 QYNTTKDKAFTDLNSINSVLGGGILDRLRPNIIPVLDEIKSMATAIKETKEALENMNSTLKSLHQQSTQLSSSLTSVKTSLRSSLNDPLCLVHPSSETCNSIRLSLS -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18245412 Leverandør Novus Biologicals™ Leverandørnr. NBP259789PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CD133 The CD133 Recombinant Protein Antigen is derived from E. coli. The CD133 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Protein længde LQDFAACGIDRMNYDSYLAQTGKSPAGVNLLSFAYDLEKANSLPPGNLRNSLKRDAQTIKTIHQQRVLPIEQSLSTLYQSVKILQRTGNGLLERVTRILASLDFAQNFITNNTSSVIIEETTKKYGRTII
Oprensningsmetode >80% by SDS-PAGE and Coomassie blue staining
Fælles navn PROM1 Recombinant Protein Antigen
Indhold og opbevaring Store at −20°C. Avoid freeze-thaw cycles.
Formulering PBS and 1M Urea, pH 7.4
Til brug med (applikation) AC
Gene Alias AC133 Stargardt disease 4 (autosomal dominant), AC133, CD133 retinal 2, CD133, PROM1
Gen symbol PROM1
Etikettype Unlabeled
Produkttype Recombinant Protein Antigen
Mængde 0.1 mL
Regulatorisk status RUO
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.