Learn More
CD133 (Prominin-1) Monoclonal Antibody (EMK08), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1230.00 DKK - 3160.00 DKK
Specifications
Antigen | CD133 (Prominin-1) |
---|---|
Clone | EMK08 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15983234
|
Affymetrix eBioscience
25-1339-42 |
100 Tests |
3160.00 DKK
Pack of 100 |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15973234
|
Affymetrix eBioscience
25-1339-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The EMK08 monoclonal antibody reacts with human CD133 (Prominin-1), a 120 kDa member of the pentaspan family of proteins, which also includes Prominin-2. Their expression is found within plasma membrane protrusions such as epithelial microvilli. CD133 can exist in a number of alternatively spliced isoforms, and the protein has several N-linked glycosylation sites; the occurrence of both may be tissue-dependent. Human CD133 was first identified as an epitope expressed on CD34+ hematopoietic progenitors. Although the ligand and function of CD133 remain unknown, it has since proven to be very useful as a marker for both stem cells and cancer stem cells. In addition to its expression on hematopoietic precursors, CD133 has been used to identify tumorigenic colon cancer stem cells, brain cancer stem cells, prostate cancer stem cells, in addition to others. The binding of the EMK08 antibody does not block the binding of another anti-human CD133 antibody, TMP4 (cat. 12-1338) indicating that they recognize distinct epitopes. Applications Reported: This EMK08 antibody has been reported for use in flow cytometric analysis. Applications Tested: This EMK08 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
CD133 (Prominin-1) Monoclonal antibody specifically detects CD133 (Prominin-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD133 (Prominin-1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O43490 | |
PROM1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
PROM1 |
EMK08 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
8842 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |