missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, anti-Human, R718, Clone: B-ly6, BD Horizon™
Mouse Monoclonal Antibody
1655.00 DKK - 3545.00 DKK
Specifications
Antigen | CD11c |
---|---|
Clone | B-ly6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17227724
|
BD Biosciences
566933 |
25 Tests |
1655.00 DKK
25 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17217724
|
BD Biosciences
566932 |
100 Tests |
3545.00 DKK
100 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
Specifications
CD11c | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
B-ly6 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |