missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD11c Antibody (N418), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81066H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for ELISA.
Tekniske data
| CD11c | |
| Monoclonal | |
| HRP | |
| PBS | |
| ITGAX | |
| Hamster spleen cells that had been primed to mouse spleen dendritic cells | |
| 0.1 mL | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Human, Mouse | |
| Purified |
| ELISA | |
| N418 | |
| ELISA | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds to mouse and human CD11c. | |
| Store at 4°C in the dark. | |
| IgG κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion