missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-Kit/SCF-Receptor Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.
1004.70 DKK - 5240.00 DKK
Specifications
Antigen | CD117/c-Kit/SCF-Receptor Ab-3 |
---|---|
Clone | K45 |
Applications | Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12680827
|
Epredia
MS-289-P1 |
200μg/mL | 500 μL |
3745.00 DKK
500µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690827
|
Epredia
MS-289-PABX |
1mg/mL | 200 μL |
5240.00 DKK
200µL |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11302824
|
Epredia
MS-289-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12600837
|
Epredia
MS-289-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12670827
|
Epredia
MS-289-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells. Ab-3 does not interfere with the binding of SCF to c-kitHost Species: Mouse
Clone: K45
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Extracellular second N-terminus Ig-like domain of c-kit
Immunogen: Recombinant human extracellular c-kit fragment
Molecular Weight: 145kDa
Positive Control: Skin, GI Stromal Tumor (GIST)
Cellular Localization: Membrane/cytoplasmic
Recommended for:
- Immunohistochemistry (Frozen or fixed in methacarn or Stieve's and embedded in paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
CD117/c-Kit/SCF-Receptor Ab-3 | |
Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
IgG2a |
K45 | |
Monoclonal | |
Concentrated | |
RUO | |
Recombinant human extracellular c-kit fragment | |
Primary |