Learn More
CD117 (c-Kit) Monoclonal Antibody (2B8), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 62-1171-80
Additional Details : Weight : 0.01000kg
Description
Description: The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145-kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. Applications Reported: This 2B8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2B8 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.
When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. c-Kit, also known as CD117 and stem cell factor receptor, is a 145 kDa transmembrane tyrosine kinase encoded by the c-Kit proto-oncogene. c-Kit acts to regulate a variety of biological responses including cell proliferation, apoptosis, chemotaxis and adhesion. Ligand binding to the extracellular domain leads to autophosphorylation on several tyrosine residues within the cytoplasmic domain, and activation. c-Kit mutations correlate with tumor growth and progression in a variety of cancers including mast cell disease, gastrointestinal stromal tumor, acute myeloid leukemia, Ewing sarcoma, and lung cancer. Phosphorylation at tyrosine 703 of c-Kit allows binding of Grb2 and activation of the Ras-Raf-ERK1&2 signaling pathway.Specifications
CD117 (c-Kit) | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q2HWD6, P05532 | |
KIT | |
Affinity chromatography | |
RUO | |
16590, 396810 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2B8 | |
Super Bright 436 | |
Kit | |
Steel Factor Receptor | |
Rat | |
25 μg | |
Primary | |
Mouse, Porcine | |
Antibody | |
IgG2b κ |
For Research Use Only.