missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD117/c-kit Antibody (rC117/9253), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23868-20ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Tekniske data
| CD117/c-kit | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA : 2-4 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Recombinant full-length human CD117/c-kit protein | |
| 20 μg | |
| B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Store at 4°C. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| rC117/9253 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion