missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ CD117/c-kit Antibody (CDX-0158), Novus Biologicals™
Human Monoclonal Antibody
1575.00 DKK - 6845.00 DKK
Tekniske data
| Antigen | CD117/c-kit |
|---|---|
| Klon | CDX-0158 |
| Applikationer | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Konjugeret | Unconjugated |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
30046124
|
Novus Biologicals™
NBP3-28628-50ug |
50 μg |
1575.00 DKK
50µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
30045110
|
Novus Biologicals™
NBP3-28628-1mg |
1 mg |
6845.00 DKK
1mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
30048707
|
Novus Biologicals™
NBP3-28628-100ug |
100 μg |
2410.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssayTekniske data
| CD117/c-kit | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| SCFR/ c-Kit/ CD117 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| CDX-0158 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P10721 | |
| 3815 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel