Learn More
CD103 (Integrin alpha E) Monoclonal Antibody (Ber-ACT8), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 17-1037-42
Description
Description: The Ber-ACT8 monoclonal antibody reacts with human CD103, the alphaE integrin. CD103 non-covalently associates with integrin beta7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. \I{In vitro\I} stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen (E-cadherin) binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium. Cross-blocking studies suggest that antibody clones B-Ly7 and Ber-ACT8 see different epitopes. Applications Reported: This Ber-ACT8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Ber-ACT8 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 ÎĽL (0.06 ÎĽg) per test. A test is defined as the amount (ÎĽg) of antibody that will stain a cell sample in a final volume of 100 ÎĽL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 ÎĽm post-manufacturing filtered.
CD103 (ITGAE, Integrin alpha E) is molecule that is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. In particular, CD103 includes a 150kDa alpha chain and a 120kDa beta chain. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is part of the integrin family which are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. ITGAE encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, CD103 protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. CD103 is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation. Diseases associated with CD103 dysfunction include hairy cell leukemia and splenic marginal zone lymphoma.
Specifications
CD103 (Integrin alpha E) | |
Monoclonal | |
5 ÎĽL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P38570 | |
Itgae | |
Affinity chromatography | |
RUO | |
3682 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
Ber-ACT8 | |
APC | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.