Få mere at vide
Tocris Bioscience™ CD 2314
GREENER_CHOICE
Artikelnummer. 30001998
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001998 10 mg
30002012 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001998 Leverandør Tocris Bioscience™ Leverandørnr. 3824/10
Selective RARβ agonist
Potent and selective RARβ receptor agonist (Kd values are 145 and >3760 nM for RARβ and RARα receptors respectively; no binding detected at RARγ). Inhibits growth of human HNSCC 22B, 183A and 22A cells (IC50 values are 3.0, 5.7 and 8.0 μM respectively).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 170355-37-0
Molekylær formel C23H24O2S
Tekniske data
Kemisk navn eller materiale CD 2314
Mængde 10 mg
Formel vægt 364.5
Fysisk form Solid
Farve White
Renhed >98%
Molekylvægt (g/mol) 364.503