Få mere at vide
CCT244747, MedChemExpress
Artikelnummer. 16463596
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
25 mg
50 mg
Pakningsstørrelse:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16463596 25 mg 25mg
16457478 10 mg -
16449957 5 mg -
16410306 2 mg -
16403507 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16463596 Leverandør Medchem Express Leverandørnr. HY18175
MedChemExpress CCT244747 is a potent, orally bioavailable and highly selective CHK1 inhibitor, with an IC50 of 7.7 nM; CCT244747 also abrogates G2 checkpoint with an IC50 of 29 nM.
CCT244747 is a potent, orally bioavailable and highly selective CHK1 inhibitor, with an IC50 of 7.7 nM
Tekniske data
Kemisk navn eller materiale CCT244747
CAS 1404095-34-6
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C20H24N8O2
Mængde 25 mg
Opløselighedsinformation DMSO : 50 mg/mL (122.41 mM; Need ultrasonic)
SMIL N#CC1=NC=C(NC2=NC=C(C3=CN(C)N=C3)C(OC)=C2)N=C1O[C@H](C)CN(C)C
Molekylvægt (g/mol) 408.46
Formel vægt 408.46
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease