Få mere at vide
CCT241736, MedChemExpress
Artikelnummer. 16479947
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16479947 5 mg -
16467468 10 mg 10mg
16437436 100 mg -
16413497 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16479947 Leverandør Medchem Express Leverandørnr. HY18161
MedChemExpress CCT241736 is a potent and orally bioavailable dual FLT3 and Aurora kinase inhibitor, which inhibits Aurora kinases (Aurora-A Kd, 7.5 nM, IC50, 38 nM; Aurora-B Kd, 48 nM), FLT3 kinase (Kd, 6.2 nM), and FLT3 mutants including FLT3-ITD (Kd, 38 nM) and FLT3(D835Y) (Kd, 14 nM).
CCT241736 is a potent and orally bioavailable dual FLT3 and Aurora kinase inhibitor, which inhibits Aurora kinases (Aurora-A Kd, 7.5 nM, IC50, 38 nM
Tekniske data
Kemisk navn eller materiale CCT241736
CAS 1402709-93-6
Farve White
Renhedsgrad noter Research
Molekylær formel C22H23Cl2N7
Mængde 5 mg
Opløselighedsinformation DMSO : 75 mg/mL (164.34 mM; Need ultrasonic)
SMIL CC1=NN(C)C=C1C2=NC3=C(N4CCN(CC5=CC=C(Cl)C=C5)CC4)C(Cl)=CN=C3N2
Molekylvægt (g/mol) 456.37
Formel vægt 456.37
Procent renhed 98.09%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease