missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Antibody (002), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89234AF405
Additional Details : Weight : 0.00970kg
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Western Blot.Specifications
CCR1 | |
Monoclonal | |
Alexa Fluor 405 | |
C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human CCR1. | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1230 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
2 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |