missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ CCKAR Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518609
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody is predicted to react with mouse and rabbit based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
Tekniske data
| CCKAR | |
| Polyclonal | |
| Unconjugated | |
| CCKAR | |
| AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 24889 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P30551 | |
| CCKAR | |
| Synthetic peptide sequence (KFDASQKKSAKEKR) corresponding to the internal amino acids of Cckar (aa 256-266). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion