missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCHC-type Zinc Finger Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-17576-25UL
2308.00 DKK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
CCHC-type Zinc Finger Polyclonal antibody specifically detects CCHC-type Zinc Finger in Human samples. It is validated for Immunofluorescence
Specifications
CCHC-type Zinc Finger | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
PNMA7B, SIZN2, ZCCHC18 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VVSLYPSLTPTGAPPFRGRARPLDQVL | |
25 μg | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
644353 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction