missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CCDC9 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05897AF647
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CCDC9 Polyclonal specifically detects CCDC9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Tekniske data
| CCDC9 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| coiled-coil domain containing 9, coiled-coil domain-containing protein 9, DKFZP586M1019 | |
| Produced in rabbits immunized with E. coli-derived Human CCDC9 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 26093 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion