Få mere at vide
Cbx8 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-38870FR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Cbx8 Polyclonal antibody specifically detects Cbx8 in Human samples. It is validated for Western Blot
Tekniske data
| DyLight 680 | |
| chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1 | |
| Primary |
| 50mM Sodium Borate | |
| Rabbit | |
| Store at 4C in the dark. |