Få mere at vide
CBR-5884, MedChemExpress
Beskrivelse
CBR-5884 is an active, selective inhibitor of phosphoglycerate dehydrogenase (PHGDH) with an IC50 of 33 μM. CBR-5884 inhibits de novo serine synthesis in cancer cells and is selectively toxic to cancer cell lines with high serine biosynthetic activity. CBR-5884 selectively inhibits the proliferation of melanoma and breast cancer lines that have a high propensity for serine synthesis.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | CBR-5884 |
| Fysisk form | Solid |
| CAS | 681159-27-3 |
| Farve | Yellow |
| Mængde | 50 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C14H12N2O4S2 |
| Til brug med (applikation) | Cancer-programmed cell death |
| Opløselighedsinformation | DMSO : 50 mg/mL (148.64 mM; Need ultrasonic) |
| SMIL | O=C(C1=C(C)C(SC#N)=C(NC(C2=CC=CO2)=O)S1)OCC |
| Vis mere |