missing translation for 'onlineSavingsMsg'
Få mere at vide

CBR-5884, MedChemExpress

Artikelnummer. 16494196
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494196 100 mg 100mg
16475477 5 mg -
16459436 50 mg -
16452738 10 mg -
16432516 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16494196 Leverandør Medchem Express Leverandørnr. HY100012

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress CBR-5884 is an active, selective inhibitor of phosphoglycerate dehydrogenase (PHGDH) with an IC50 of 33 μM. CBR-5884 inhibits de novo serine synthesis in cancer cells and is selectively toxic to cancer cell lines with high serine biosynthetic activity. CBR-5884 selectively inhibits the proliferation of melanoma and breast cancer lines that have a high propensity for serine synthesis.

CBR-5884 is an active, selective inhibitor of phosphoglycerate dehydrogenase (PHGDH) with an IC50 of 33 μM. CBR-5884 inhibits de novo serine synthesis in cancer cells and is selectively toxic to cancer cell lines with high serine biosynthetic activity. CBR-5884 selectively inhibits the proliferation of melanoma and breast cancer lines that have a high propensity for serine synthesis.

Tekniske data

Kemisk navn eller materiale CBR-5884
Fysisk form Solid
CAS 681159-27-3
Farve Yellow
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C14H12N2O4S2
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : 50 mg/mL (148.64 mM; Need ultrasonic)
SMIL O=C(C1=C(C)C(SC#N)=C(NC(C2=CC=CO2)=O)S1)OCC
Molekylvægt (g/mol) 336.39
Formel vægt 336.39
Procent renhed 98.74%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.