missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Caveolin-1 Antibody (7C8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80617
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Tekniske data
| Caveolin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:1000 - 1:4000, Flow Cytometry : 1 ug per million cells, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence : 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin : 1:100 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774) | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 7C8 | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| CAV1 | |
| Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes | |
| 0.1 mL | |
| Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
| 857 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion