Learn More
Minigrip™ Resealable Write-on Bags
Product Code. 10001044
Change view
Click to view available options
Width (Metric):
40 mm
75 mm
100 mm
140 mm
190 mm
Height (Metric):
65 mm
85 mm
140 mm
190 mm
Unit Size:
Pack of 100
Pack of 1000
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
10001044 140 mm 140 mm Pack of 100
11326924 75 mm 85 mm Pack of 100
10205074 100 mm 140 mm Pack of 100
10366984 190 mm 190 mm Pack of 100
10634185 40 mm 65 mm Pack of 100
10758235 75 mm 190 mm Pack of 100
30057395 75 mm 190 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10001044 Supplier Minigrip™ Supplier No. BAJ340060B
  • ONLINE EXCLUSIVE PRICE
    75.44 DKK
    23.65 DKK / Pakke med 100
    Spare 51.79 DKK
    68% Slukket
Forventet forsendelse: 04-01-2027
Label bag contents with ease. Minigrip™ Resealable Write-on Bags provide three white bars specifically designed for writing on with inkpen or marker.
  • Polyethylene
  • Resealable
  • Autoclavable
  • Three write-on bars for labelling
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Height (Metric) 140 mm
Width (Metric) 140 mm