missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99088AF647
Additional Details : Weight : 0.00970kg
Description
Cathepsin G Polyclonal specifically detects Cathepsin G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cathepsin G | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cathepsin G, CathepsinG, CGCATG, EC 3.4.21, EC 3.4.21.20, MGC23078 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Cathepsin G. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
1511 | |
Store at 4°C in the dark. | |
IgG |