Få mere at vide
Cathepsin E Antibody (215), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90637G
Produktkode 18310811
  • 4670.00 DKK / 0.10ml
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Cathepsin E
Monoclonal
DyLight 488
CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin E (NP_031825.2; Met1-Pro397).
0.1 mL
Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism
1510
Mouse
Purified
ELISA
215
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Mouse CTSB Mouse CTSC
Store at 4°C in the dark.
IgG