missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Cathepsin B Recombinant Rabbit Monoclonal Antibody (6I5D7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535194
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: SDELVNYVNK RNTTWQAGHN FYNVDMSYLK RLCGTFLGGP KPPQRVMFTE DLKLPASFDA REQWPQCPTI KEIRDQGSCG SCWAFGAVEA ISDRICIHTN AHVSVEVSAE DLLTCCGSMC GDGCNGGYPA.
Cathepsin B is a gene located on chromosome 8p23.1 that encodes a lysosomal cysteine protease involved in intracellular protein degradation. As a member of the cathepsin family, Cathepsin B plays a crucial role in the lysosomal pathway, facilitating the breakdown of proteins into amino acids, which can then be recycled for cellular functions. In addition to its proteolytic activity within lysosomes, Cathepsin B is also implicated in extracellular matrix remodeling, cellular invasion, and apoptotic pathways. Its expression and activity are often upregulated in various cancer types, including breast, colon, and pancreatic cancers, where it contributes to tumor progression and metastasis by degrading extracellular matrix components and promoting tumor cell invasion. Due to these roles, Cathepsin B is considered a potential biomarker for cancer prognosis and a target for therapeutic intervention aimed at inhibiting cancer cell dissemination. Research continues to explore its broader implications in diseases related to protease activity and inflammation.
Tekniske data
| Cathepsin B | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide, pH 7.3 | |
| P07858 | |
| Ctsb | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6I5D7 | |
| Unconjugated | |
| Ctsb | |
| amyloid precursor protein secretase; APP secretase; APPS; cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; CB; CPSB; Ctsb; cysteine protease; RSG-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1508 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion