missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cathepsin B Inhibitor IV
Click to view available options
Quantity:
1 mg
10 mg
Description
A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Specifications
Quantity | 1 mg |
Description | White |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B. |
Formula Weight | 397.46g/mol |
Solubility | Soluble in DMSO |
Product Type | Signal Transduction Reagent |
Form | Powder |
InChI Key | XGWSRLSPWIEMLQ-UHFFFAOYNA-N |
SMILES | CCCNC(=O)C1OC1C(=O)NC(C(C)CC)C(=O)N1CCCC1C(=O)OC |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'