missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Cathelicidin Recombinant Rabbit Monoclonal Antibody (K01_1N95)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

3644.00 DKK - 3715.00 DKK

Tekniske data

Antigen Cathelicidin
Klon K01_1N95
Koncentration 1 mg/mL
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Applikationer Western Blot
Se flere specifikationer

Produkter 2
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
30826152
GREENER_CHOICE
Dokumentvisning
Invitrogen™
MA563280
100 μL
3644.00 DKK
100µL
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
30828953
Dokumentvisning
Invitrogen™
MA564739
50 μL
3715.00 DKK
50µL
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

Cathelicidin Recombinant Monoclonal Antibody for Western Blot

Cathelicidin is an epithelial-derived antimicrobial protein found in specific granules of polymorphonuclear leukocytes of bone marrow, testis and neutrophils. Cathelicidins are one family of antimicrobial peptides characterized by conserved pro-peptide sequences that have been identified in epithelial tissues and some myeloid cells of humans and animals. LL-37/hCAP-18 is the only Cathelicidin found in humans and is expressed in inflammatory and epithelial cells. The presence of these molecules is essential for defense against invasive bacterial infection in skin. Besides their direct antimicrobial function, Cathelicidins have multiple roles in mediating innate and adaptive immunity, such as endotoxin neutralizing, angiogenesis, wound healing and promoting neutrophil chemotaxis and mast cell recruitment. Cathelicidin antimicrobial peptides qualify as prototypes of innovative drugs that may be used to treat infection and/or modulate the immune response.
Tekniske data

Tekniske data

Cathelicidin
1 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
P49913
820
A synthetic peptide of human Cathelicidin.
Antibody
K01_1N95
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Recombinant Monoclonal
Liquid
IgG
Human
CAMP
18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; Camp; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; CRAMP; FALL39; FALL-39; FALL-39 peptide antibiotic; hCAP18; hCAP-18; HSD26; LL37; MCLP
CAMP
Primary
Protein A/G
Videoer
SDS
Dokumenter

Dokumenter

Produktcertifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.