Få mere at vide
Catharanthine Tartrate, TRC
Artikelnummer. 30366209
Skift visning
Mængde:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366209 1 g
30360396 10 g
30361805 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366209 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2254001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4168-17-6
Molekylær formel 2 C21 H24 N2 O2 . C4 H6 O6
Molekylvægt (g/mol) 822.94
IUPAC navn 2,3-dihydroxybutanedioic acid;methyl (1R,15R,18R)-17-ethyl-3,13-diazapentacyclo[13.3.1.02,10.04,9.013,18]nonadeca-2(10),4,6,8,16-pentaene-1-carboxylate
SMIL CCC1=C[C@@H]2CN3CCc4c([nH]c5ccccc45)[C@@](C2)([C@@H]13)C(=O)OC.CCC6=C[C@@H]7CN8CCc9c([nH]c%10ccccc9%10)[C@@](C7)([C@@H]68)C(=O)OC.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Catharanthine Tartrate
Mængde 1 g
InChI formel InChI=1S/2C21H24N2O2.C4H6O6/c2*1-3-14-10-13-11-21(20(24)25-2)18-16(8-9-23(12-13)19(14)21)15-6-4-5-7-17(15)22-18;5-1(3(7)8)2(6)4(9)10/h2*4-7,10,13,19,22H,3,8-9,11-12H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t2*13-,19+,21-;1-,2-/m001/s1
Formel vægt 822.384