Få mere at vide
Catechol-d6, TRC
Artikelnummer. 30357225
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30357225 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30357225 Leverandør Toronto Research Chemicals Leverandørnr. TRCC215178500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Catechol-d6
CAS 202656-22-2
Renhedsgrad noter HPLC
Molekylær formel C6 2H6 O2
Mængde 500 mg
Synonym 1,2-Benzene-3,4,5,6-d4-diol-d2, Pyrogallol-d6, 1,2-Benzenediol-1,2,3,4,5,6-D6, Catechol-1,2,3,4,5,6-D6, Catechol D6, Benzene-1,2-diol D6
InChI formel InChI=1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H/i1D,2D,3D,4D/hD2
SMIL [2H]Oc1c([2H])c([2H])c([2H])c([2H])c1O[2H]
IUPAC navn 1,2,3,4-tetradeuterio-5,6-dideuteriooxybenzene
Molekylvægt (g/mol) 116.15
Formel vægt 116.0744
Procent renhed >95