Få mere at vide
(+/-)-Catechin xHydrate, TRC
Artikelnummer. 30132593
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132593 500 mg
30134914 1 g
30007796 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132593 Leverandør Toronto Research Chemicals Leverandørnr. TRCC217520500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7295-85-4
Molekylvægt (g/mol) 290.27
IUPAC navn (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMIL O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3
Tekniske data
Kemisk navn eller materiale DL-Catechin
Molekylær formel C15 H14 O6
Mængde 500 mg
Synonym 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-(±)-, Catechol, (±)- (8CI), rel-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (±)-Catechin, (±)-Catechol, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-, DL-Catechin, Racemic catechin, dl-Catechin, dl-Catechol, rac-Catechin
InChI formel InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1
Formel vægt 290.079