Få mere at vide
(±)-Catechin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | DL-Catechin |
| CAS | 7295-85-4 |
| Analyt- eller komponentnavne | DL-Catechin |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H14 O6 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | (2R,3S)-rel-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (±)-Catechin, (±)-Catechol, rac-Catechin, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-(±)-, Catechol, (±)- (8CI), rel-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (±)-Catechin, (±)-Catechol, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-, DL-Catechin, Racemic catechin, dl-Catechin, dl-Catechol, rac-Catechin |
| InChI formel | InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1 |
| SMIL | O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3 |
| Vis mere |