missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Catalyst Norprene™ Food Formulation Pump Tubing
Available in standard 15m Lengths.
455.00 DKK - 975.00 DKK
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
15541305
|
Fisherbrand™
|
1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15551305
|
Fisherbrand™
|
3.2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15561305
|
Fisherbrand™
|
4.8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15571305
|
Fisherbrand™
|
6.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15581305
|
Fisherbrand™
|
8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
- Available in both thin and thick wall configurations
Specifications
Catalyst pump tubing | |
1.6 mm |