missing translation for 'onlineSavingsMsg'
Learn More
Casticin, MedChemExpress Product Code.: 16443876

Casticin, MedChemExpress

Product Code. p-200098126
25 mg, 25mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16443876

Brand: Medchem Express HYN0516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Casticin is a methyoxylated flavonol isolated from Viticis Fructus, with antimitotic and anti-inflammatory effect. Casticin inhibits the activation of STAT3.

Casticin is a methyoxylated flavonol isolated from Viticis Fructus, with antimitotic and anti-inflammatory effect. Casticin inhibits the activation of STAT3.

Specifications

Chemical Name or Material Casticin
CAS 479-91-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H18O8
Quantity 25 mg
Synonym Vitexicarpin
Solubility Information DMSO : 100 mg/mL (267.14 mM; Need ultrasonic)
SMILES O=C1C(OC)=C(C2=CC=C(OC)C(O)=C2)OC3=CC(OC)=C(OC)C(O)=C13
Molecular Weight (g/mol) 374.34
Formula Weight 374.34
Percent Purity 98.4%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Casticin, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.