Få mere at vide
Invitrogen™ CaspGLOW™ Fluorescein Active Caspase-9 Staining Kit
Mængde:
100 tests
Pakningsstørrelse:
100 tests
Beskrivelse
Indeholder
- FITC-LEHD-FMK
- Wash Buffer
- Z-VAD-FMK
- Uses the inhibitor specific for Caspase-9, Fluorescein (FITC)-conjugated LEHD-FMK, for detection
- This reagent is cell permeable, non-toxic and irreversibly binds to the active enzyme
Tekniske data
Tekniske data
| Mængde | 100 tests |
| Format | Kit |
| Produkttype | Staining Kit |
| Til brug med (applikation) | Flow Cytometry |
| Til brug med (udstyr) | Fluorescence Microscope, Flow Cytometer |
For Research Use Only. Not for use in diagnostic procedures.