missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Caspase-8 Inhibitor I, cell-permeable
Brand: Alfa Aesar™ J66050.|D
Additional Details : Weight : 0.00001kg
Specifications
0.5mg | |
−30° to −10°C | |
C95H162N20O26 | |
N-Acetyl-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Ile-Glu-Thr-Asp-al; Ac-AAVALLPAVLLALLAPIETD-CHO | |
Lyophilized powder |
White to Off-white | |
Lyophilized Powder | |
2000.41g/mol | |
Signal Transduction Reagent |