missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Caspase-3 Substrate III, fluorogenic

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66881.M¬

Additional Details : CAS Number : 189696-20-6 Weight : 0.00001kg

 View more versions of this product

Product Code. 15478419

  • 1812.28 DKK / 1mg


Estimated Shipment
28-10-2020
Add to basket

Specifications

Specifications

Fluorogenic
-30° to -10°C
Lyophilized
Lyophilized powder
1mg
White to off-white
1155.08
Videos
SDS Documents

For Research Use Only