missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CASK, GST Tag Recombinant Protein

CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. Recombinant human CASK (1-570) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Brand:  Invitrogen™ PV6273

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15408933

  • 55700.00 DKK / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
91.0kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
1mg
aa 1-570
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers