missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein, Hammarsten Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 lb.
100 g
11.3 kg (25 lb.)
2.2 kg (5 lb.)
Description
- Cream to light yellow
- Protein: ≥95%
- Melting point: ∽280°C (dec.)
- MDLNumber: MFCD00081481
- CAS: 9000-71-9
Specifications
Molecular Weight (g/mol) | 75000 to 100000 |
Name | Casein |
Quantity | 2.2 kg (5 lb.) |
Packaging | Plastic bottle |
Sensitivity | Hygroscopic |
Form | Lyophilized |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'