missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein, Hammarsten Grade, Thermo Scientific Chemicals
Cream to light yellow, lyophilized powder.
279.84 DKK - 11379.10 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15845098
|
Thermo Scientific
J12840.22 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15855098
|
Thermo Scientific
J12840.TD |
1 lb. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15865098
|
Thermo Scientific Alfa Aesar
J12840.P5 |
2.2 kg (5 lb.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15875098
|
Thermo Scientific
J12840.5 |
11.3 kg (25 lb.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Cream to light yellow
- Protein: ≥95%
- Melting point: ∽280°C (dec.)
- MDLNumber: MFCD00081481
- CAS: 9000-71-9
RUO – Research Use Only