Få mere at vide
Carvedilol EP Impurity D, Mikromol™
Artikelnummer. 30462461
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462461 4 x 25 mg
30051066 100 mg
30050944 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462461 Leverandør Mikromol Leverandørnr. MM0291.1300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Carvedilol EP Impurity D, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[(2-(2-methoxyphenoxy)ethyl)amino]propoxy]-9H-carbazol-9-yl]propan-2-ol
CAS 1391052-16-6
Analyt- eller komponentnavne 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[(2-(2-methoxyphenoxy)ethyl)amino]propoxy]-9H-carbazol-9-yl]propan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C39 H39 N3 O6
Certificeringer/compliance ISO 9001
Synonym 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[[2-(2-methoxyphenoxy)ethyl]amino]propoxy]-9H-carbazol-9-yl]propan-2-ol, 9H-Carbazole-9-ethanol, alpha-[(9H-carbazol-4-yloxy)methyl]-4-[2-hydroxy-3-[[2-(2-methoxyphenoxy)ethyl]amino]propoxy]-, Carvedilol Impurity D
InChI formel InChI=1S/C39H39N3O6/c1-45-34-16-6-7-17-35(34)46-21-20-40-22-26(43)24-47-37-19-9-15-33-39(37)29-11-3-5-14-32(29)42(33)23-27(44)25-48-36-18-8-13-31-38(36)28-10-2-4-12-30(28)41-31/h2-19,26-27,40-41,43-44H,20-25H2,1H3
Løsningstype Neat
SMIL COc1ccccc1OCCNCC(O)COc2cccc3c2c4ccccc4n3CC(O)COc5cccc6[nH]c7ccccc7c56
IUPAC navn 1-[9-[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]carbazol-4-yl]oxy-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
Molekylvægt (g/mol) 645.74
Mængde 4 x 25 mg
Formel vægt 645.2839 g/mol