Få mere at vide
Carvedilol-d5, TRC
Artikelnummer. 30352154
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30352154 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352154 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1846281MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carvedilol-d5
CAS 929106-58-1
Renhedsgrad noter HPLC
Molekylær formel C24H21D5N2O4
Mængde 1 mg
Synonym 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol-d5, Artist-d5, BM 14190, Cadilan-d5, Carca-d5, Cardivas-d5, Carloc-d5, Carvas-d5, Carvedilol-d5, Carvediol-d5, Coreg-d5, DQ 2466-d5, Dilatrend-d5, Dimitone-d5, Eucardic-d5, Korvasan-d5, Kredex-d5, Querto-d5, SKF 105517-d5, Talliton-d5
InChI formel InChI=1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3/i15D2,16D2,17D
SMIL OC(C([2H])([2H])NCCOC1=C(OC)C=CC=C1)([2H])C([2H])([2H])OC2=CC=CC3=C2C4=C(N3)C=CC=C4
IUPAC navn 1-(9H-carbazol-4-yloxy)-1,1,2,3,3-pentadeuterio-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
Molekylvægt (g/mol) 411.51
Procent renhed >95